missing translation for 'onlineSavingsMsg'
Learn More

2-Benzothiazolemethanol, 97%, Thermo Scientific Chemicals

£34.67 - £117.59

Chemical Identifiers

CAS 37859-42-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00226293
InChI Key PQXMQZYDBQBWNL-UHFFFAOYSA-N
Synonym benzo d thiazol-2-ylmethanol, 2-benzothiazolemethanol, 2-hydroxymethylbenzothiazole, 1,3-benzothiazol-2-yl-methanol, benzothiazol-2-ylmethan-1-ol, zlchem 107, pubchem14535, benzothiazol-2-yl-methanol, chembl71700, 1,3-benzothiazol-2-yl methanol
PubChem CID 268122
IUPAC Name 1,3-benzothiazol-2-ylmethanol
SMILES OCC1=NC2=CC=CC=C2S1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494227
View Documents
Thermo Scientific Alfa Aesar
H31631.03
1 g
£34.67
1g
Please to purchase this item. Need a web account? Register with us today!
15404237
View Documents
Thermo Scientific Alfa Aesar
H31631.06
5 g
£117.59
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37859-42-0
165.21
PQXMQZYDBQBWNL-UHFFFAOYSA-N
268122
OCC1=NC2=CC=CC=C2S1
C8H7NOS
MFCD00226293
benzo d thiazol-2-ylmethanol, 2-benzothiazolemethanol, 2-hydroxymethylbenzothiazole, 1,3-benzothiazol-2-yl-methanol, benzothiazol-2-ylmethan-1-ol, zlchem 107, pubchem14535, benzothiazol-2-yl-methanol, chembl71700, 1,3-benzothiazol-2-yl methanol
1,3-benzothiazol-2-ylmethanol

Specifications

37859-42-0
C8H7NOS
benzo d thiazol-2-ylmethanol, 2-benzothiazolemethanol, 2-hydroxymethylbenzothiazole, 1,3-benzothiazol-2-yl-methanol, benzothiazol-2-ylmethan-1-ol, zlchem 107, pubchem14535, benzothiazol-2-yl-methanol, chembl71700, 1,3-benzothiazol-2-yl methanol
OCC1=NC2=CC=CC=C2S1
165.21
165.21
2-Benzothiazolemethanol
96°C to 100°C
MFCD00226293
PQXMQZYDBQBWNL-UHFFFAOYSA-N
1,3-benzothiazol-2-ylmethanol
268122
97%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only