missing translation for 'onlineSavingsMsg'
Learn More

2-Benzothiazolemethanol, 97%, Thermo Scientific Chemicals

Product Code. 15404237
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37859-42-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00226293
InChI Key PQXMQZYDBQBWNL-UHFFFAOYSA-N
Synonym benzo d thiazol-2-ylmethanol, 2-benzothiazolemethanol, 2-hydroxymethylbenzothiazole, 1,3-benzothiazol-2-yl-methanol, benzothiazol-2-ylmethan-1-ol, zlchem 107, pubchem14535, benzothiazol-2-yl-methanol, chembl71700, 1,3-benzothiazol-2-yl methanol
PubChem CID 268122
IUPAC Name 1,3-benzothiazol-2-ylmethanol
SMILES OCC1=NC2=CC=CC=C2S1

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material 2-Benzothiazolemethanol
compliance-icons
Product Identifier
  • 2-Benzothiazolemethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hydroxymethylbenzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'