Learn More
2',4'-Dimethylacetanilide, 98%, Thermo Scientific™
£26.20 - £108.00
Chemical Identifiers
CAS | 2050-43-3 |
---|---|
Molecular Formula | C10H13NO |
Molecular Weight (g/mol) | 163.22 |
MDL Number | MFCD00026125 |
InChI Key | PJEIIBXJUDOMAN-UHFFFAOYSA-N |
Synonym | n-2,4-dimethylphenyl acetamide, 2',4'-dimethylacetanilide, 2,4-dimethylacetanilide, 4-acetamido-m-xylene, 2',4'-acetoxylidine, 2',4'-acetoxylidide, acetamide, n-2,4-dimethylphenyl, acetanilide, 2',4'-dimethyl, n-acetyl-2,4-xylidine, unii-3u0w52hz1h |
PubChem CID | 16303 |
IUPAC Name | N-(2,4-dimethylphenyl)acetamide |
SMILES | CC1=CC(=C(C=C1)NC(=O)C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2050-43-3 | |
163.22 | |
PJEIIBXJUDOMAN-UHFFFAOYSA-N | |
16303 | |
CC1=CC(=C(C=C1)NC(=O)C)C |
C10H13NO | |
MFCD00026125 | |
n-2,4-dimethylphenyl acetamide, 2',4'-dimethylacetanilide, 2,4-dimethylacetanilide, 4-acetamido-m-xylene, 2',4'-acetoxylidine, 2',4'-acetoxylidide, acetamide, n-2,4-dimethylphenyl, acetanilide, 2',4'-dimethyl, n-acetyl-2,4-xylidine, unii-3u0w52hz1h | |
N-(2,4-dimethylphenyl)acetamide |
Specifications
2050-43-3 | |
White | |
MFCD00026125 | |
PJEIIBXJUDOMAN-UHFFFAOYSA-N | |
N-(2,4-dimethylphenyl)acetamide | |
16303 | |
98% | |
2',4'-Dimethylacetanilide |
129°C to 132°C | |
C10H13NO | |
n-2,4-dimethylphenyl acetamide, 2',4'-dimethylacetanilide, 2,4-dimethylacetanilide, 4-acetamido-m-xylene, 2',4'-acetoxylidine, 2',4'-acetoxylidide, acetamide, n-2,4-dimethylphenyl, acetanilide, 2',4'-dimethyl, n-acetyl-2,4-xylidine, unii-3u0w52hz1h | |
CC1=CC(=C(C=C1)NC(=O)C)C | |
163.22 | |
163.22 | |
Powder |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 218-088-6
missing translation for 'rtecsNumber' : AN7420000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures