missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Melting Point | ∽240°C (decomposition) |
Assay Percent Range | 0.99 |
Quantity | 5 g |
UN Number | UN1325 |
Formula Weight | 164.11 |
Percent Purity | 97% |
Chemical Name or Material | 1H-Tetrazole-5-carboxylic acid ethyl ester sodium salt |
Safety and Handling
Product Identifier
- 1H-Tetrazole-5-carboxylic acid ethyl ester,sodium salt
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'