missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyne, 99%, Thermo Scientific Chemicals

Catalog Number p-7043746
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-19-0
C5H8
68.12
MFCD00009469
IBXNCJKFFQIKKY-UHFFFAOYSA-N
1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
12309
pent-1-yne
CCCC#C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-19-0
C5H8
68.12
MFCD00009469
IBXNCJKFFQIKKY-UHFFFAOYSA-N
1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
12309
pent-1-yne
CCCC#C

1-Pentyne has been used in preparation of:lithium acetylides, required for asymmetric synthesis of α,α-dibranched propargyl sulfinamides. It is also used to synthesize 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentyne has been used in preparation of:lithium acetylides, required for asymmetric synthesis of α,α-dibranched propargyl sulfinamides. It is also used to synthesize 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans.

Solubility
Soluble in water. (1.05 g/L) at 20°C.

Notes
Store at 2-8°C. Incompatible with oxidizing agents. Store away from heat.
TRUSTED_SUSTAINABILITY
Melting Point -106°C
Density 0.692
Boiling Point 39°C to 41°C
Flash Point −20°C (−4°F)
Odor Unpleasant
Linear Formula CH3CH2CH2C≡CH
Refractive Index 1.386
Quantity 5 g
UN Number UN3295
Beilstein 1697133
Solubility Information Soluble in water. (1.05g/L) at 20°C.
Formula Weight 68.12
Percent Purity 99%
Chemical Name or Material 1-Pentyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'