missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Catalog Number p-3736097
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
13419-61-9
C10H21NaO3S
244.32
MFCD00007526
AIMUHNZKNFEZSN-UHFFFAOYSA-M
sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
2724181
sodium;decane-1-sulfonate
CCCCCCCCCCS(=O)(=O)[O-].[Na+]
13419-61-9
C10H21NaO3S
244.32
MFCD00007526
AIMUHNZKNFEZSN-UHFFFAOYSA-M
sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
2724181
sodium;decane-1-sulfonate
CCCCCCCCCCS(=O)(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY
Absorbance 0.02 max. (C=0.2 M in water, 40°C) at 230nm, 0.02 max. (C=0.2 M in water, 40°C) at 260nm, 0.03 max. (C=0.2 M in water, 40°C) at 220nm, 0.05 max. (C=0.2 M in water, 40°C) at 210nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 1% max. (150°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)9SO3Na
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 244.32
Percent Purity 99+%
Grade Ion Pair Chromatography
Physical Form Crystalline Powder
Chemical Name or Material 1-Decanesulfonic acid, sodium salt, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'