Learn More
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%, Thermo Scientific Chemicals
£31.76 - £332.00
Chemical Identifiers
CAS | 762240-99-3 |
---|---|
Molecular Formula | C4H4ClF3N2O2 |
Molecular Weight (g/mol) | 204.53 |
MDL Number | MFCD07779449 |
InChI Key | DYKIVKLXFDNBMY-UHFFFAOYSA-N |
Synonym | 1-chloroacetyl-2-trifluoroacetyl hydrazine, n'-2-chloroacetyl-2,2,2-trifluoroacetohydrazide, n'-chloroacetyl-2,2,2-trifluoroacetohydrazide, acetic acid, 2,2,2-trifluoro-, 2-2-chloroacetyl hydrazide, 2-chloro-n'-2,2,2-trifluoroacetyl acetohydrazide, n'-2-chloroacetyl-trifluoroacetohydrazide, 1-trifluoroacetyl-2-chloroacetyl hydrazine, 2-chloro-n'-trifluoroacetyl acetohydrazide, acetic acid, trifluoro-, 2-chloroacetyl hydrazide |
PubChem CID | 11961370 |
IUPAC Name | N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide |
SMILES | FC(F)(F)C(=O)NNC(=O)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15451436
|
Thermo Scientific Alfa Aesar
H54647.MD |
250 mg |
£31.76
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15490878
|
Thermo Scientific Alfa Aesar
H54647.03 |
1 g |
£90.05
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400888
|
Thermo Scientific Alfa Aesar
H54647.06 |
5 g |
£332.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
762240-99-3 | |
204.53 | |
DYKIVKLXFDNBMY-UHFFFAOYSA-N | |
11961370 | |
FC(F)(F)C(=O)NNC(=O)CCl |
C4H4ClF3N2O2 | |
MFCD07779449 | |
1-chloroacetyl-2-trifluoroacetyl hydrazine, n'-2-chloroacetyl-2,2,2-trifluoroacetohydrazide, n'-chloroacetyl-2,2,2-trifluoroacetohydrazide, acetic acid, 2,2,2-trifluoro-, 2-2-chloroacetyl hydrazide, 2-chloro-n'-2,2,2-trifluoroacetyl acetohydrazide, n'-2-chloroacetyl-trifluoroacetohydrazide, 1-trifluoroacetyl-2-chloroacetyl hydrazine, 2-chloro-n'-trifluoroacetyl acetohydrazide, acetic acid, trifluoro-, 2-chloroacetyl hydrazide | |
N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide |
Specifications
762240-99-3 | |
MFCD07779449 | |
DYKIVKLXFDNBMY-UHFFFAOYSA-N | |
N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide | |
11961370 | |
97% |
C4H4ClF3N2O2 | |
1-chloroacetyl-2-trifluoroacetyl hydrazine, n'-2-chloroacetyl-2,2,2-trifluoroacetohydrazide, n'-chloroacetyl-2,2,2-trifluoroacetohydrazide, acetic acid, 2,2,2-trifluoro-, 2-2-chloroacetyl hydrazide, 2-chloro-n'-2,2,2-trifluoroacetyl acetohydrazide, n'-2-chloroacetyl-trifluoroacetohydrazide, 1-trifluoroacetyl-2-chloroacetyl hydrazine, 2-chloro-n'-trifluoroacetyl acetohydrazide, acetic acid, trifluoro-, 2-chloroacetyl hydrazide | |
FC(F)(F)C(=O)NNC(=O)CCl | |
204.53 | |
204.53 | |
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only