missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%, Thermo Scientific Chemicals

Product Code. 15400888
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762240-99-3
Molecular Formula C4H4ClF3N2O2
Molecular Weight (g/mol) 204.53
MDL Number MFCD07779449
InChI Key DYKIVKLXFDNBMY-UHFFFAOYSA-N
Synonym 1-chloroacetyl-2-trifluoroacetyl hydrazine, n'-2-chloroacetyl-2,2,2-trifluoroacetohydrazide, n'-chloroacetyl-2,2,2-trifluoroacetohydrazide, acetic acid, 2,2,2-trifluoro-, 2-2-chloroacetyl hydrazide, 2-chloro-n'-2,2,2-trifluoroacetyl acetohydrazide, n'-2-chloroacetyl-trifluoroacetohydrazide, 1-trifluoroacetyl-2-chloroacetyl hydrazine, 2-chloro-n'-trifluoroacetyl acetohydrazide, acetic acid, trifluoro-, 2-chloroacetyl hydrazide
PubChem CID 11961370
IUPAC Name N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide
SMILES FC(F)(F)C(=O)NNC(=O)CCl

Specifications

Quantity 5 g
Formula Weight 204.53
Percent Purity 97%
Chemical Name or Material 1-Chloroacetyl-2-(trifluoroacetyl)hydrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'