Learn More
1-Bromoundecane, 98%, Thermo Scientific Chemicals
£29.15 - £84.22
Chemical Identifiers
CAS | 693-67-4 |
---|---|
MDL Number | MFCD00000223 |
InChI Key | IKPSIIAXIDAQLG-UHFFFAOYSA-N |
Synonym | undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane |
PubChem CID | 12744 |
IUPAC Name | 1-bromoundecane |
SMILES | CCCCCCCCCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582811
|
Thermo Scientific Acros
224010250 |
25 g | Glass bottle |
£29.15
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10184510
|
Thermo Scientific Acros
224011000 |
100 g | Glass bottle |
£84.22
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
693-67-4 | |
IKPSIIAXIDAQLG-UHFFFAOYSA-N | |
12744 | |
CCCCCCCCCCCBr |
MFCD00000223 | |
undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane | |
1-bromoundecane |
Specifications
693-67-4 | |
100.0 | |
Colorless to Yellow | |
137°C to 138°C (18.0 mmHg) | |
Authentic | |
C11H23Br | |
1.4555 to 1.4575 | |
1.05 | |
Solubility in water: insoluble | |
CCCCCCCCCCCBr | |
235.21 | |
235.21 | |
Liquid |
97.5 | |
-9°C | |
1.0500g/mL | |
>110°C | |
97.5% min. (GC) | |
CH3(CH2)10Br | |
MFCD00000223 | |
undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane | |
IKPSIIAXIDAQLG-UHFFFAOYSA-N | |
1-bromoundecane | |
12744 | |
98% | |
1-Bromoundecane, 99% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-757-3
missing translation for 'tsca' : TSCA
RUO – Research Use Only