missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-cyclohexylbenzene, 97%, Thermo Scientific Chemicals

Product Code. 11324895
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59734-92-8
Molecular Formula C12H15Br
Molecular Weight (g/mol) 239.16
MDL Number MFCD02093987
InChI Key BXMDAGGQRIDSPA-UHFFFAOYSA-N
Synonym 1-bromo-2-cyclohexyl-benzene, benzene, 1-bromo-2-cyclohexyl, acmc-20aoxa, l-bromo-2-cyclohexylbenzene, 2-bromo-1-cyclohexylbenzene
PubChem CID 10944502
IUPAC Name 1-bromo-2-cyclohexylbenzene
SMILES BrC1=CC=CC=C1C1CCCCC1

Specifications

Density 1.295
Boiling Point 79°C to 83°C (0.01 mmHg)
Refractive Index 1.5605
Quantity 5 g
Beilstein 2365764
Formula Weight 239.16
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-cyclohexylbenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'