missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%, Thermo Scientific Chemicals
£27.50 - £90.30
Chemical Identifiers
CAS | 17938-13-5 |
---|---|
Molecular Formula | C16H22Si2 |
Molecular Weight (g/mol) | 270.522 |
MDL Number | MFCD00135433 |
InChI Key | CMTMWEXUJQSPCA-UHFFFAOYSA-N |
Synonym | 1,4-bis trimethylsilyl ethynyl benzene, 1,4-bis trimethylsilylethynyl benzene, 1,4-bis 2-trimethylsilyl ethynyl benzene, trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane, trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane, acmc-20ap2w, trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane, benzene,1,4-bis 2-trimethylsilyl ethynyl |
PubChem CID | 522492 |
IUPAC Name | trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane |
SMILES | C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312227
|
Thermo Scientific Alfa Aesar
L09709.03 |
1 g |
£27.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322227
|
Thermo Scientific Alfa Aesar
L09709.06 |
5 g |
£90.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17938-13-5 | |
270.522 | |
CMTMWEXUJQSPCA-UHFFFAOYSA-N | |
522492 | |
C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C |
C16H22Si2 | |
MFCD00135433 | |
1,4-bis trimethylsilyl ethynyl benzene, 1,4-bis trimethylsilylethynyl benzene, 1,4-bis 2-trimethylsilyl ethynyl benzene, trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane, trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane, acmc-20ap2w, trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane, benzene,1,4-bis 2-trimethylsilyl ethynyl | |
trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane |
Specifications
119°C to 121°C | |
C16H22Si2 | |
2840618 | |
CMTMWEXUJQSPCA-UHFFFAOYSA-N | |
trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane | |
522492 | |
98% |
17938-13-5 | |
MFCD00135433 | |
1,4-bis trimethylsilyl ethynyl benzene, 1,4-bis trimethylsilylethynyl benzene, 1,4-bis 2-trimethylsilyl ethynyl benzene, trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane, trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane, acmc-20ap2w, trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane, benzene,1,4-bis 2-trimethylsilyl ethynyl | |
C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C | |
270.522 | |
270.53 | |
1,4-Bis[(trimethylsilyl)ethynyl]benzene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only