missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyrimidinyl)homopiperazine, Thermo Scientific Chemicals

Product Code. 11479280
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21279-57-2
Molecular Formula C9H14N4
Molecular Weight (g/mol) 178.24
MDL Number MFCD06738258
InChI Key LZOGPVCTSDAYIP-UHFFFAOYSA-N
Synonym 1-pyrimidin-2-yl-1,4-diazepane, 1-2-pyrimidinyl homopiperazine, 1-pyrimidin-2-yl-1,4 diazepane, 1-pyrimidin-2-yl-1,4-diazaperhydroepine, acmc-20aorx, 1-2-pyrimidyl homopiperazine, 1-2-pyrimidyl-homopiperazine
PubChem CID 2772409
IUPAC Name 1-pyrimidin-2-yl-1,4-diazepane
SMILES C1CNCCN(C1)C1=NC=CC=N1

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 178.24
Chemical Name or Material 1-(2-Pyrimidinyl)homopiperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'