missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)ethane nickel(II) chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-3777525
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14647-23-5
C26H24Cl2NiP2
528.02
MFCD00013313
XXECWTBMGGXMKP-UHFFFAOYSA-L
1,2-bis diphenylphosphino ethane nickel ii chloride
131664338
nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
14647-23-5
C26H24Cl2NiP2
528.02
MFCD00013313
XXECWTBMGGXMKP-UHFFFAOYSA-L
1,2-bis diphenylphosphino ethane nickel ii chloride
131664338
nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride
Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 263.0°C to 265.0°C
Color Orange to Red
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 528.02
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 2-Bis(diphenylphosphino)ethane nickel(II) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'