missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dichlorophosphino)ethane, 97%, Thermo Scientific Chemicals

Catalog Number p-67694
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
28240-69-9
C2H4Cl4P2
231.81
MFCD00009609
SBWAJHLQMFBNIN-UHFFFAOYSA-N
1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472
119904
dichloro(2-dichlorophosphanylethyl)phosphane
C(CP(Cl)Cl)P(Cl)Cl
This item is not returnable. View return policy
28240-69-9
C2H4Cl4P2
231.81
MFCD00009609
SBWAJHLQMFBNIN-UHFFFAOYSA-N
1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472
119904
dichloro(2-dichlorophosphanylethyl)phosphane
C(CP(Cl)Cl)P(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Density 1.5300g/mL
Boiling Point 70.0°C (1.0 mmHg)
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula Cl2PCH2CH2PCl2
Refractive Index 1.5875 to 1.5935
Quantity 1 g
Specific Gravity 1.53
Solubility Information Solubility in water: reacts
Formula Weight 231.81
Percent Purity 97%
Chemical Name or Material 1, 2-Bis(dichlorophosphino)ethane, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'