missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ White Mixed Cellulose Esters Sterile Gridded S-Pak Filters

Brand:  Millipore™ RAWG047S6

Code : NEW

Additional Details : Weight : 0.12000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II

Product Code. 11963856

  • £422.00 / Pack of 600


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

White
Gridded
600Pack
For Microbiological analysis
1.2μm
47mm
S-Pak Filters
Sterile
Mixed Cellulose Ester
S-Pak Filters
Videos
Documents