missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Precolumn Inline Filters

Protect larger columns and injections from harmful particulates

Brand:  Idex A-318

Code : A0

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11574835

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Ideal for higher-flow systems
  • Interchangeable frits make protection more economical
  • Compatible with SFC and HPLC
  • Available to fit a variety of tubing sizes
Specifications

Specifications

0.5 μm
Precolumn Inline Filter
1 in.
Stainless Steel
0.23 uL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers