Learn More
Thermo Scientific™ TopVision Agarose PROMO
TopVision Agarose is highly purified agarose suitable for analytical and preparative electrophoresis of nucleic acids.
Brand: Thermo Scientific™ R0491-RMD123
Additional Details : Weight : 0.18000kg
Description
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.Specifications
Agarose Gel |