missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gene Frame, 65 µL (1.5 x 1.6 cm)

Gene Frames for in-situ feature a dual strength adhesive system which facilitates the easy removal of the frames after use.

£79.75 - £90.25


Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11550294 View Documents Thermo Scientific™
AB0576
25μL £79.75
Pack of 100 View alternative products
Save Up to 
»
EU Stock 48
Add to basket
 
11560294 View Documents Thermo Scientific™
AB0577
65μL £85.75
Pack of 100 View alternative products
Save Up to 
»
EU Stock 2
Add to basket
 
11570294 View Documents Thermo Scientific™
AB0578
125μL £90.25
Pack of 100 View alternative products
Save Up to 
»
EU Stock 14
Add to basket
 
Description

Description

Thermo Scientific Gene Frames have adhesives on either side of the frame which are of differing strengths; the side which adheres to the glass has an easy release adhesive whereas the reverse, which sticks to the coverslip, forms a stronger bond. This means that when the coverslip is removed, due to the disparity in the strengths of the different adhesives, the frame is removed simultaneously with no sticky residue being left behind on the slide.

Highlights
  • Gas tight sealing prevents reagent loss due to evaporation
  • Can withstand temperatures of 97°C
  • Designed for use with standard microscope slides
  • Frame and coverslip can be removed simultaneously leaving no residue
  • Can be autoclaved or sterilized by UV irradiation
  • All Gene Frame packs are supplied with 100 coverslips
  • Rigid coverslips for ease of handling and uniform hybridizations
  • Thickness of Gene Frames: 0.25 mm
Applications
  • In-situ PCR

Hazardous: No
Storage Condition: Ambient

Certifications

Certified free from DNase, RNase and human genomic DNA.