missing translation for 'onlineSavingsMsg'
Learn More

BD Syringes

Brand:  BD 300300

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 12319299

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Opaque
107.5 mm
1 mL
30 mL
Sterile
23.77 mm
Syringe
240/Pk.
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers