Learn More
Thermo Scientific™ SuperFrost Ultra Plus™ Adhesion Slides
The surface of SuperFrost Ultra Plus© slides is positively charged, but is characterized by optimized tissue adhesion when in-situ hybridization techniques or immunoperoxidase procedures with heat-induced antigen/epitope retrieval (HIER, HMAR or HTAR) are required. For tissue slices 2 to 5 micrometers thick.
£32.65 - £59.75
Specifications
Length (Metric) 75mm
Width (Metric) 25mm
Thickness (Metric) 1mm
View More Specs
Products 2
Product Code Brand Edges Price Quantity & Availability  
Product Code Brand Edges Price Quantity & Availability  
10417002 View Documents Thermo Scientific™
J3800AMNZ
90° This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10235612 View Documents Thermo Scientific™
J4800AMNZ
45° This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • No time wasted on slide-coating proceduress
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
Specifications
Specifications
75mm
25mm
1mm