missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamide, 99%, Thermo Scientific Chemicals
41.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
2.5 g
10 g
50 g
Description
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.
Solubility
Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol
Notes
Moisture Sensitive. Store away from oxidizing agents, water/humidity. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.
Solubility
Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol
Notes
Moisture Sensitive. Store away from oxidizing agents, water/humidity. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 91°C to 92°C |
Density | 1.61 |
Quantity | 10 g |
Sensitivity | Moisture sensitive |
Merck Index | 14,8922 |
Solubility Information | Soluble in water, hot alcohol, acetone; Sparingly soluble in cold alcohol |
IUPAC Name | sulfamoylamine |
Formula Weight | 96.11 |
Percent Purity | 99% |
Chemical Name or Material | Sulfamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'