missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Storage Vials & Closures

Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.

Brand:  Thermo Scientific™ B7990-2A

Code : 17

Additional Details : Weight : 0.68000kg

 View more versions of this product

Product Code. 12438666

  • £39.95 / Pack of 100


Estimated Shipment
07-09-2020
Add to basket

Description

Description

  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.

Unassembled convenience kits saves time during sample preparation.

  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Specifications

Specifications

Screw Cap (Open-Top)
Amber
4 mL
Screw Vial Convenience Kit, Open top Cap
Glass
100
Videos
Documents

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.