Learn More
Sonics Materials™ Stainless Steel Optional Temperature Probe
Brand: Sonics Materials™
Code : A0
Additional Details : Weight : 0.07000kg
Specifications
Optional Temperature Probe | |
Allows sample temperature to be monitored up to 100°C | |
1 Ea. |
Stainless Steel | |
VCX |