missing translation for 'onlineSavingsMsg'
Learn More

SPE accessory reservoirs for use with 3mL and 6mL columns 75mL

Brand:  7120-03

SPE accessory reservoirs for use with 3mL and 6mL columns 75mL

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 13107584

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.