Learn More
Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I, Thermo Scientific™
Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds
Brand:  Thermo Scientific Alfa Aesar 044893.22
Code : NEW
Additional Details : Weight : 0.10000kg Transport : UN number : 2813 Chem class : 4.3 Pack group : II
 View more versions of this product
Product Code. 15476625
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Sodium-silica gel
Black
100 g
Reacts with water.
0.7
35-40% alkali metal in silica gel, Stage I
Powder
MFCD01941718
Moisture sensitive
Product Suggestions
Product Suggestions
Videos
SDS Documents