missing translation for 'onlineSavingsMsg'
Learn More

Sodium deuteroxide, 40% w/w solution in D{2}O, 99.5%(Isotopic), Thermo Scientific Chemicals

Catalog Number p-4407880
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
14014-06-3
HNaO
41.003
MFCD00037669
HEMHJVSKTPXQMS-DYCDLGHISA-M
sodium deuteroxide, sodium hydroxide na od, sodium deuteroxide, 40 wt% in deuterium oxide, naod, sodium hydroxide-d, sodium deuteroxide solution, sodium deuteroxide, 1 m in deuterium oxide, sodium deuteroxide, 0.1 m in deuterium oxide, sodium deuteroxide, 30 wt% in deuterium oxide, sodium hydroxide na od 6ci,7ci,8ci,9ci
23676750
[OH-].[Na+]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
14014-06-3
HNaO
41.003
MFCD00037669
HEMHJVSKTPXQMS-DYCDLGHISA-M
sodium deuteroxide, sodium hydroxide na od, sodium deuteroxide, 40 wt% in deuterium oxide, naod, sodium hydroxide-d, sodium deuteroxide solution, sodium deuteroxide, 1 m in deuterium oxide, sodium deuteroxide, 0.1 m in deuterium oxide, sodium deuteroxide, 30 wt% in deuterium oxide, sodium hydroxide na od 6ci,7ci,8ci,9ci
23676750
[OH-].[Na+]

Sodium deuteroxide is a reagent used in the synthesis of rac Zearalenone-d6. is used in many chemical reactions and industrial practices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Quantity 10 g
Assay Percent Range 99.5%
Solubility Information Fully miscible in water.
Formula Weight 41.01
Odor Odorless
Refractive Index 1.418
Sensitivity Air and moisture sensitive
Appearance Colorless liquid
Density 1.516
Chemical Name or Material Sodium deuteroxide, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'