Learn More
Thermo Scientific™ SLIPFREE™ Sample Loops
Thermo Scientific™ SLIPFREE Sample Loops feature a self-adjusting, leak-free design.
£96.25 - £158.00
Specifications
Type Sample Loop
For Use With Liquid Chromatography
View More Specs
Products 5
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11568710 View Documents Thermo Scientific™
31620
10 µL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10418104 View Documents Thermo Scientific™
31640
20 µL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10661026 View Documents Thermo Scientific™
32625
50 µL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10741967 View Documents Thermo Scientific™
32650
100 µL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10187704 View Documents Thermo Scientific™
32699
250 µL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Compatible with Rheodyne and Valco™ injectors
  • Long neck design
Specifications
Specifications
Sample Loop
Liquid Chromatography