missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ 3.5K MWCO Dialysis Cassettes

Remove buffer salts and contaminants and maximize sample recovery using this semi-permeable membrane with a 3500-dalton protein MW cut-off (MWCO).

£88.25 - £155.00

Specifications

MWCO 3500 Da
View More Specs

Products 7
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10416144 View Documents Thermo Scientific™
66107
Slide-A-Lyzer Dialysis Cassette Kit, 3.5K MWCO, 12mL £155.00
Each View alternative products
Save Up to 
»
EU Stock 7 Please to purchase this item. Need a web account? Register with us today!
10500814 View Documents Thermo Scientific™
66110
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 12mL £142.00
Pack of 8 View alternative products
Save Up to 
»
EU Stock 97 Please to purchase this item. Need a web account? Register with us today!
10501004 View Documents Thermo Scientific™
66130
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 30mL £88.25
Pack of 6 View alternative products
Save Up to 
»
EU Stock 40 Please to purchase this item. Need a web account? Register with us today!
10759784 View Documents Thermo Scientific™
66330
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 3mL £123.00
Pack of 10 View alternative products
Save Up to 
»
EU Stock 119 Please to purchase this item. Need a web account? Register with us today!
10751565 View Documents Thermo Scientific™
66332
Slide-A-Lyzer Dialysis Cassette Kit, 3.5K MWCO, 3mL £146.00
Each View alternative products
Save Up to 
»
EU Stock 12 Please to purchase this item. Need a web account? Register with us today!
11859410 View Documents Thermo Scientific™
66333
Slide-A-Lyzer Dialysis Cassettes, 3.5K MWCO, 0.5mL £121.00
Pack of 10 View alternative products
Save Up to 
»
EU Stock 20 Please to purchase this item. Need a web account? Register with us today!
10455685 View Documents Thermo Scientific™
66335
Slide-A-Lyzer Dialysis Cassette Kit, 3.5K MWCO, 0.5mL £141.00
Each View alternative products
Save Up to 
»
EU Stock 12 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Available in four different cassette sizes to accommodate sample volumes between 0.5 and 30mL, these devices are easier to use than traditional dialysis tubing, providing both faster buffer exchange and more complete sample recovery. Slide-A-Lyzer Cassettes can be used for a wide range of applications including low molecular-weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.

The cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Highlights:

  • Easy to use – no knots or clamps are needed; just inject sample into cassette and begin dialysis

  • Fast dialysis – flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing

  • High recovery – rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports

  • Four cassette sizes – select the cassette that best suits your sample volume

  • Color-coded – pink cassette frame specifies cassettes with the 3500 MW pore size membrane

3.5K Dialysis Membrane Specifications:

  • Composition: Regenerated cellulose synthesized by the Viscose method
  • Thickness: 0.9 to 1.2mil (22.5 to 30μm)
  • Hydration Required Before Use: 30 seconds
  • Glycerol Content: Trace
  • Sulfur Content: 0.1 to 0.15%
  • Heavy Metals Content: Trace

Item-Specific Features:

    30mL cassette has built-in air chamber for flotation

Includes:

Kits contain cassettes plus an equal number of syringes with 18-gauge needles and single-cassette float buoys

Specifications

Specifications

3500 Da