missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SKF-96365 hydrochloride, 99+%

Catalog Number p-7082250
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
130495-35-1
C22H26N2O3·HCl, C22H27ClN2O3
1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole
This item is not returnable. View return policy
130495-35-1
C22H26N2O3·HCl, C22H27ClN2O3
1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole

Inhibits receptor-mediated calcium influx

Inhibits receptor-mediated calcium influx

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 mg
Description White
For Use With (Application) Inhibits receptor-mediated calcium influx
Formula Weight 402.92g/mol
Solubility Soluble to 100mM in water
Product Type Signal Transduction Reagent
Form Powder
InChI Key FWLPKVQUECFKSW-UHFFFAOYNA-N
SMILES [H+].[Cl-].COC1=CC=C(CCCOC(CN2C=CN=C2)C2=CC=C(OC)C=C2)C=C1
IUPAC Name hydrogen 1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole chloride
Molecular Weight (g/mol) 402.92
Assay ≥99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'