Learn More
Thermo Scientific™ Shelving Options and Accessories for Environmental Chambers and Incubators
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
£233.00 - £366.00
Specifications
Material Stainless Steel
Type Shelf
View More Specs
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15503771 View Documents Thermo Scientific™
224139
Solid Stainless Steel Shelf and Channels £233.00
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
15513771 View Documents Thermo Scientific™
224155
Perforated Stainless Steel Shelf and Channels £348.31
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
15523771 View Documents Thermo Scientific™
224161
Reinforced Stainless Steel Shelf and Channels (max. load 68kg) £366.00
Each View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
11851130 View Documents Thermo Scientific™
224161
Reinforced Stainless Steel Shelf and Channels (max. load 68kg) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Specifications
Specifications
Stainless Steel
Shelf
Cat No: 224161 -- Two per unit max., not for shakers; 68.0 kg load maximum with shelf fully inserted and stationary