Learn More
Sharpsafe™ Impenetrable Polypropylene Disposal Containers
Use Sharpsafe™ Impenetrable Polypropylene Disposal Containers for incinerations or alternative treatments and for outstanding, impenetrable protection from leaks or sharps.
POA
Specifications
Material | Polypropylene |
---|---|
Certifications/Compliance | Manufactured to BS 7320: 1990, ISO 9002 standards and UN Packing Group II approved |
Model | Rectangular |
Quantity | 1 Ea. |
Includes | Closure |
Description
- Yellow with orange lid
- Clear British Standard markings
- Visible fill line
Specifications
Polypropylene | |
Rectangular | |
Closure |
Manufactured to BS 7320: 1990, ISO 9002 standards and UN Packing Group II approved | |
1 Ea. |