Learn More
Sartorius Microsart™ Maxi Vacuum Pumps
Microsart Maxi vacuum pumps provide up-to-date technology for daily use in the microbiology laboratory environment
Brand:  Sartorius 16694-1-60-22
Additional Details : Weight : 9.52900kg
Product Code. 15962540
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Ideal for microbiological filtrations
  • Neoprene membrane pumps
  • Decreased sub lethals, higher recovery rates and shorter incubation times
  • Low noise level
  • Oil-and maintenance-free
  • Controlled and easily adjustable vacuum
  • Pressure filtration with Microsart mini vac possible
Specifications
Specifications
15.65 lb.
0.9 A
8 x 10.3 x 4.3 in. (204 x 261 x 110 mm)
130 w
40°C
60 Hz
10.92 cm
20.32 cm
261 mm
115 V
7.1 kg
115 V 60 Hz
57.5 to 59.0 dB (A)
104°F
22 L/min.
4.3 in.
8 in.
10.3 in.
1 bar