Learn More
Sartorius™ Extraction Thimbles
POA
Specifications
Bottom Shape Round
Product Type Extraction Thimble
Sterility Non-sterile
View More Specs
Description
Description
  • Manufactured from cotton linters with a high alpha-cellulose content
  • High-purity thimbles that possess mechanical strength and outstanding retention capacity
  • Thermally stable up to 100°C
  • Inner diameter, outer length, round bottom and wall thickness defined according to the norm DIN 12449
  • Especially suited for all routine extractions: fat extractions, extraction of lipids from eggs and egg-containing products, softeners, additives, monomer components in plastics and rubber
Specifications
Specifications
Round
Extraction Thimble
Non-sterile