missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Digital Round Tissue Section Bath

Obtain accuracy, reliability, and ease-of-use with the Epredia™ Digital Round Tissue Section Bath.

Brand:  Epredia™ A84600060

Additional Details : Weight : 4.50000kg

 View more versions of this product

Product Code. 15553646

  • £1450.00 / Each
Estimated Shipment: 26-04-2024
to see stock.
Add to basket

Description

Description

Features
  • Deep bowl for easier section pick up
  • Wide bowl accommodates longer section ribbons
  • Large area to rest hands when floating sections
  • Miniature circuit breakers for ease-of-use; simply push to reset
  • Anti-microbial paint finish (silver ion) to inhibit bacterial growth


Heating Efficiency
  • Ultra-fast heating system
  • Significant slide drying area at the rear of the instrument
  • A secure-fitting lid reduces airborne contamination, evaporation, and improves heating efficiency
  • Automatic over-temperature cut-out for safety
Specifications

Specifications

3.5 L
Certificate of Analysis (COA), Certificate of Conformance (COC)
Digital
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers