missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Septa for 8mm Screw-Thread Vials

Red PTFE

£37.20 - £277.20

Specifications

Diameter (Metric) Outer 8 mm
Material PTFE, Silicone
View More Specs

Products 4
Product Code Brand Color Quantity Price Quantity & Availability  
Product Code Brand Color Quantity Price Quantity & Availability  
13136872
View Documents Promotion Details
Restek™
21147
Red, White 1000
£277.20
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
 
13176912
View Documents Promotion Details
Restek™
21178
Red 100
£37.20
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 08-05-2024
to see stock.
Add to basket
 
13186912
View Documents Promotion Details
Restek™
21179
Red 1000
£247.20
Pack of 1000
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
 
13169682
View Documents Promotion Details
Restek™
21795
Red, White 100
£39.60
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
 
Description

Description

Minimize bleed with preconditioned, low-bleed septa.

Handle septa carefully to prevent contamination

Specifications

Specifications

8 mm
PTFE, Silicone
Videos
SDS
Documents

Documents

Product Certifications