Learn More
Eppendorf™ Replacement Lid
Use this replacement rotor lid with Rotor F-45-24-11. Eppendorf™ Replacement Lid is not aerosol-tight.
Brand:  Eppendorf™ 5425701004
Code : 99
Additional Details : Weight : 0.06000kg
Product Code. 10516911
  • £31.27 / Each
In stock, ready to ship: 13
Description
Description
  • Polypropylene lid
  • For Rotor F-45-24-11
Specifications
Specifications
Replacement lid
Standard Rotor F 45-24-11
Polypropylene