missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat (Wistar) Cryopreserved Hepatocytes, Suspension Female
Rat (Wistar) Cryopreserved Hepatocytes, Suspension Female
Brand: Gibco™ WICS20
Code : 50
Additional Details : Weight : 0.01000kg
Description
These cryopreserved female rat (Wistar) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.Cells per vial: 4–8 million
Get the Results You Need with GIBCO™ Hepatocytes
High quality hepatocytes with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure that you receive the highest quality cryopreserved hepatocytes.
Each lot is tested for:
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Links:
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
Specifications
135°C | |
Cryopreserved | |
Hepatocytes | |
Suspension Female Rat (Wistar) Cryopreserved Hepatocytes, 1mL, Rat Species, Hepatocytes Cell Type, Pooled Donor, Liquid Nitrogen Storage, For Hepatocytes and Hepatic Models, Pharma and Biopharma, Primary Cell Culture, Target-Based ADME/Tox Assays |
Hepatocytes | |
Cell culture, drug discovery and development, target-based ADME/Tox assays, cellular toxicology assays, primary cell culture, hepatocyte culture, hepatocytes and hepatic models | |
1mL |
For Research Use Only. Not for use in diagnostic procedures.