missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PTFE Tubing Kit
Includes tubing, mouting with ferrule and nut
Brand: Cytiva 28-9809-87
Code : GX
Additional Details : Weight : 0.34200kg
Description
Inlet tubing for the ÅKTA avant 150 system.Specifications
Tubing Kit | |
2.9 mm | |
10 x Tubing, mounted with one ferrule and nµt | |
1500 mm |
4.6 mm | |
1 Each | |
ÄKTA Avant 150 | |
PTFE |