missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ BPC Bag with 4 Top Ports

Using these BPC bags with 4 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

£138.00 - £990.00

Specifications

Description Three-dimensional disposable cell culture/bioprocess bag
Film Description CX3-9 Film
Port Detail 3/8in. Quick Disconnect Insert (MPC Series); 1/4in. Male Luer Lock; 1/4in. Female Luer Lock; 3/8in. Quick Disconnect Body (MPC Series)
Sterilization Method Gamma Irradiated
Storage Requirements Ambient
View More Specs

Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15566829 View Documents Thermo Scientific™
SH30651.01
50L £770.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15586829 View Documents Thermo Scientific™
SH30651.03
200L £905.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10135903 View accessory products View Documents Thermo Scientific™
SH30651.02
100L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
15576829 View Documents Thermo Scientific™
SH30651.02
100L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10014733 View accessory products View Documents Thermo Scientific™
SH30651.03
200L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10638574 View accessory products View Documents Thermo Scientific™
SH30651.01
50L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

  • Square tube design, 3-dimensional

  • Constructed with Thermo Scientific CX5-14 film with a contact surface composed of ultra-low density polyethylene
  • Four top ports (2 to 25.4mm (1in.); 2 to 12.7mm (1/2in.))
  • Line 1:

  • 9.5mm (3/8in.) MPC insert constructed of polycarbonate with 121.9cm (48in.) of C-Flex™ SEBS tubing (9.5mm (3/8in.) ID by 15.9mm
  • (5/8in.) OD); Variable length diptube: 50L - 38cm (15in.); 100L - 71cm (28in.); 200L - 81cm (32in.)
  • Line 2:

  • 6.4mm (1/4in.) Male Luer Lock constructed of polypropylene with 121.9cm (48in.) of C-Flex SEBS tubing (6.4mm (1/4in.) ID by 9.5mm (3/8in.) OD)
  • Line 3:

  • 6.4mm (1/4in.) Female Luer Lock constructed of polypropylene with 121.9cm (48in.) of C-Flex SEBS tubing (6.4mm (1/4in.) ID by 9.5mm (3/8in.) OD)
  • Line 4:

  • 9.5mm (3/8in.) MPC body constructed of polycarbonate with 121.9cm (48in.) of C-Flex SEBS tubing (9.5mm (3/8in.) ID by 15.9mm (5/8in.) OD);
  • Diptube length:
  • 30.5cm (12in.)
  • Designed to be used with flat bottom linear low density polyethylene drums with clamps and matching lids
  • Plastic drum dolly for ergonomic convenience
Specifications

Specifications

Three-dimensional disposable cell culture/bioprocess bag
3/8in. Quick Disconnect Insert (MPC Series); 1/4in. Male Luer Lock; 1/4in. Female Luer Lock; 3/8in. Quick Disconnect Body (MPC Series)
Ambient
CX3-9 Film
Gamma Irradiated
No