missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ BPC Bag with 2 Top Ports

Using these BPC bags with 2 top ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

£117.00 - £1405.00

Specifications

Description Three-dimensional disposable cell culture/bioprocess bag; 100L; Two Top Ports: MPC Insert, MPC Body with 48 in. Dip Tube and 12 in. Dip Tube
Film Description CX3-9 Film
Port Detail 3/8in. Quick Disconnect Insert (MPC Series); 3/8in. Quick Disconnect Body (MPC Series)
Sterilization Method Gamma Irradiated
Storage Requirements Ambient
View More Specs

Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15506829 View Documents Thermo Scientific™
SH30649.01
50L £1030.00
Pack of 8 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15516829 View Documents Thermo Scientific™
SH30649.02
100L £1405.00
Pack of 10 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15526829 View Documents Thermo Scientific™
SH30649.03
200L £785.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10004443 View accessory products View Documents Thermo Scientific™
SH30649.01
50L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10752634 View accessory products View Documents Thermo Scientific™
SH30649.03
200L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10247753 View accessory products View Documents Thermo Scientific™
SH30649.02
100L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainer BPCs can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

Three-dimensional disposable cell culture/bioprocess bag; 100L; Two Top Ports: MPC Insert, MPC Body with 48 in. Dip Tube and 12 in. Dip Tube
3/8in. Quick Disconnect Insert (MPC Series); 3/8in. Quick Disconnect Body (MPC Series)
Ambient
CX3-9 Film
Gamma Irradiated
No