missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ BPC Bag with 2 Top and 1 Bottom Ports

Using these BPC bags with 2 top and 1 bottom ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

£132.00 - £1200.00

Specifications

Description Three-dimensional disposable cell culture/bioprocess bag; 100L; Tri-Clamp Port.(Bottom)
Film Description CX5-14 Film
Sterilization Method Gamma Irradiated
Storage Requirements Ambient
View More Specs

Products 12
Product Code Brand Port Detail Capacity (Metric) Price Quantity & Availability  
Product Code Brand Port Detail Capacity (Metric) Price Quantity & Availability  
15536829 View Documents Thermo Scientific™
SH30650.01
1/2in. Quick Disconnect Insert (MPX Series); 1/2in. Quick Disconnect Body (MPX Series); 1/4in. Female Luer Lock 50L £880.00
Pack of 6 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15546829 View Documents Thermo Scientific™
SH30650.02
1/2in. Quick Disconnect Insert (MPX Series); 1/2in. Quick Disconnect Body (MPX Series); 1/4in. Female Luer Lock 100L £805.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15556829 View Documents Thermo Scientific™
SH30650.03
1/2in. Quick Disconnect Insert (MPX Series); 1/2in. Quick Disconnect Body (MPX Series); 1/4in. Female Luer Lock 200L £865.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15586839 View Documents Thermo Scientific™
SH30672.01
3/4in (Mini) TriClamp Connector x 3 50L £1180.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15596839 View Documents Thermo Scientific™
SH30672.02
3/4in (Mini) TriClamp Connector x 3 100L £920.00
Pack of 4 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
15506849 View Documents Thermo Scientific™
SH30672.03
3/4in (Mini) TriClamp Connector x 3 200L £1200.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10196513 View Documents Thermo Scientific™
SH30672.01
3/4in (Mini) TriClamp Connector x 3 50L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10024873 View accessory products View Documents Thermo Scientific™
SH30650.01
1/2in. Quick Disconnect Insert (MPX Series); 1/2in. Quick Disconnect Body (MPX Series); 1/4in. Female Luer Lock 50L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10369913 View Documents Thermo Scientific™
SH30672.02
3/4in (Mini) TriClamp Connector x 3 100L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10218333 View accessory products View Documents Thermo Scientific™
SH30650.03
1/2in. Quick Disconnect Insert (MPX Series); 1/2in. Quick Disconnect Body (MPX Series); 1/4in. Female Luer Lock 200L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10064483 View Documents Thermo Scientific™
SH30672.03
3/4in (Mini) TriClamp Connector x 3 200L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
10106143 View accessory products View Documents Thermo Scientific™
SH30650.02
1/2in. Quick Disconnect Insert (MPX Series); 1/2in. Quick Disconnect Body (MPX Series); 1/4in. Female Luer Lock 100L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

Productainer - 2 Top/1 Bottom Ports:

  • Square tube design, 3-dimensional
  • Constructed with Thermo Scientific CX 5-14 film with a contact surface composed of ultra-low density polyethylene
  • Two top ports (12.7mm (1/2in.) and 6.4mm (1/4in.))
  • Line 1: 12.7mm (1/2in.) MPX Body constructed of polycarbonate with 45.7cm (18in.) of C-Flex™ SEBS tubing (12.7mm (1/2in.) ID by 19.1mm (3/4in.) OD)
  • Line 2: 6.4mm (1/4in.) Female Luer Lock constructed of polypropylene with 30.5cm (12in.) of C-Flex SEBS tubing (6.4mm (1/4in.) ID by 9.5mm (3/8in.) OD)
  • One bottom port (12.7mm (1/2in.))
  • Line 3: 12.7mm (1/2in.) MPX Insert constructed of polycarbonate with 121.9cm (48in.) of C-Flex SEBS tubing (12.7mm (1/2in.) ID by 19.1mm (3/4in.) OD)
  • Designed to be used with linear low density polyethylene drums with conical bottom, and clamps and matching lids
  • Plastic drum dolly for ergonomic convenience

Productainer - 2 Top/1 Bottom Ports, Tri-Clamps:

  • Square tube design, 3-dimensional
  • Constructed with Thermo Scientific CX5-14 film with a contact surface composed of ultra-low density polyethylene
  • Two top ports (2 - 6.4mm (1/2 in.))
  • Line 1: 6.4mm (1/2 in.) Tri-Clamp (mini) constructed of polyethylene with 45.7cm (18 in.) of C-Flex SEBS tubing (6.4mm (1/2 in.) ID by 19.1mm (3/4 in.) OD)
  • Line 2: 6.4mm (1/2 in.) Tri-Clamp (mini) constructed of polyethylene with 45.7cm (18 in.) of C-Flex SEBS tubing (6.4mm (1/2 in.) ID by 19.1mm (3/4 in.) OD)
  • One bottom port (6.4mm (1/2 in.)
  • Line 3: 6.4mm (1/2 in.) Tri-Clamp (mini) constructed of polyethylene with 121.9cm (48 in.) of C-Flex SEBS tubing (6.4mm (1/2 in.) ID by 19.1mm (3/4 in.) OD)
  • Designed to be used with linear low density polyethylene drums with conical bottom, and clamps and matching lids
  • Plastic drum dolly for ergonomic convenience

Specifications

Specifications

Three-dimensional disposable cell culture/bioprocess bag; 100L; Tri-Clamp Port.(Bottom)
CX5-14 Film
Gamma Irradiated
Ambient