missing translation for 'onlineSavingsMsg'
Learn More

Potassium thioglycolate, 40-44 wt.% solution in water, ACROS Organics™

Brand:  Acros Organics 384700010

Code : 29

Additional Details : CAS Number : 34452-51-2 Weight : 1.30000kg Transport : UN number : 2922 Chem class : 8 Pack group : III


Packaging Glass bottle
Quantity 1L

 View more versions of this product

Product Code. 10638424

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

34452-51-2
130.202
HYTYHTSMCRDHIM-UHFFFAOYSA-M
23688957
C(C(=O)[O-])S.[K+]
C2H3KO2S
MFCD00067265
potassium thioglycolate, unii-xp5wzo5w6h, potassium 2-mercaptoacetate, acetic acid, mercapto-, monopotassium salt, xp5wzo5w6h, 68-11-1 parent, potassium sulfanylacetate, potassium mercaptoacetate, kopcolate, potassium ion thioglycolate
potassium;2-sulfanylacetate

Specifications

Potassium thioglycolate
7732-18-5
MFCD00067265
potassium thioglycolate, unii-xp5wzo5w6h, potassium 2-mercaptoacetate, acetic acid, mercapto-, monopotassium salt, xp5wzo5w6h, 68-11-1 parent, potassium sulfanylacetate, potassium mercaptoacetate, kopcolate, potassium ion thioglycolate
C(C(=O)[O-])S.[K+]
130.202
130.21
40.0 to 44.0%
0.5ppm max.
Solubility in water: soluble.
Pink
1L
40-44 wt.% solution in water
C2H3KO2S
1.2500g/mL
HYTYHTSMCRDHIM-UHFFFAOYSA-M
potassium;2-sulfanylacetate
23688957
Solution
potassium: positive, mercaptane: positive
Glass bottle
1.25
6.8 to 7.3
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium thioglycolate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

SDS Documents