Learn More
Equip Labo™ Polypropylene Laboratory Fume Hood
Rich, robust, reliable laboratory fume hood made of polypropylene material.
POA
Specifications
Product Type | Laboratory Fume Hood |
---|---|
Lighting Type | Glare Fluorescent Tube |
Material | Polypropylene |
Quantity | 1 Ea. |
Description
- 180 x 80 x 155 cm dimensions
- Back panel in polypropylene more chemically resistant
- Gutter for the recovery of condensates to be connected to the grid
- discharge
- Fume hood made of polypropylene material
Specifications
Laboratory Fume Hood | |
Glare Fluorescent Tube | |
Polypropylene | |
1 Ea. |