missing translation for 'onlineSavingsMsg'
Learn More

Waste bag green plain 457mm x 737 x 990mm

Brand:  LDGS

Waste bag green plain 457mm x 737 x 990mm

Code : 03

Additional Details : Weight : 0.20000kg

Product Code. 13162380

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »