Learn More
Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Other Instruments
Get accurate and reproducible results over a range of volumes with Thermo Scientific™ Plunger-in-Needle Autosampler Syringes for Other Instruments.
Brand:  Thermo Scientific™ 365A0331
Code : 16
Additional Details : Weight : 0.10000kg
Product Code. 10219364
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
Specifications
Specifications
0.5uL volume; 23 gauge; 50mm length