Learn More
Thermo Scientific™ Pierce™ High Capacity Copper Coated Plates
Analyze polyhistidine-tagged fusion proteins by ELISA with these high-capacity plates treated to maximize histidine-tagged protein binding.
£218.00 - £248.00
Specifications
Binding Target His tag
Quantity 5 Plates
Sufficient For Binding approx. 35pmol His-tagged protein per well
View More Specs
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10034413 View Documents Thermo Scientific™
15143
Pierce Copper Coated High Capacity Plates, Clear, 96-Well £231.00
Pack of 5 View alternative products
Save Up to 
»
EU Stock 7
Add to basket
10388593 View Documents Thermo Scientific™
15146
Pierce Copper Coated High Capacity Plates, Clear, 96-Well Strip £248.00
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-03-2020
Add to basket
10257623 View Documents Thermo Scientific™
15147
Pierce Copper Coated High Capacity Plates, White, 96-Well £218.00
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
10083883 View Documents Thermo Scientific™
15148
Pierce Copper Coated High Capacity Plates, Black, 96-Well £218.00
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
Description
Description
Pierce Copper Coated Plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper. These copper-coated plates are available as clear, white or black plates that can be used with colorimetric, chemiluminescent or fluorescent detection methods, respectively. Although copper is less discriminating in its binding specificity than Nickel, the higher binding capacity of these plates is ideal for high throughput screening applications that need improved sensitivity and greater dynamic range.
Highlights:
  • Quantitate previously undetectable histidine-tagged proteins
  • Wider dynamic range with four-fold greater capacity than regular nickel chelate-coated plate
  • Copper chelate provides greater binding capacity than nickel
  • Detection limit is 0.1pg of polyhistidine tagged fusion protein per well
  • Pre-blocked for the ultimate convenience
Specifications
Specifications
His tag
5 Plates
Binding approx. 35pmol His-tagged protein per well