Learn More
Invitrogen™ STAT1 (Phospho) [pY701] Human InstantOne™ ELISA Kit
InstantOne ELISA
Brand: Invitrogen™ 85-86092-11
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: User Manual
InstantOne™ Assay Plate - 96 well (sealed)
Cell Lysis Buffer (5X)
Enhancer Solution
Wash Buffer (10X)
phospho-STAT1 (Tyr701) Capture Antibody Reagent
phospho-STAT1 (Tyr701) Detection Antibody Reagent
Positive Control Cell Lysate (lyophilized)
Detection Reagent (TMB Substrate)
Stop Solution
Plate Seal
Description
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human STAT1 (Tyr701) in cell lysates. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately two hours. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
STAT1 (signal transducers and activators of transcription 1) is a member of the STAT family of transcription factors. STAT1 can be activated by interferon-alpha, interferon-gamma, EGF, PDGF and IL6. STAT1 is known to regulate several genes which are involved in cell growth, apoptosis, immune responses, and lipid metabolism. Further, STAT1 plays an important role in mediating cell viability in response to different cell stimuli and pathogen exposure. The STAT1 gene is located on chromosome 2. STAT1 is activated to regulate gene expression in response to extracellular signaling polypeptides including cytokines, interferons, and growth factors. After phosphorylation by JAK tyrosine kinases, STAT1 enters the nucleus to regulate transcription of many different genes. Among the seven STATs types, STAT1, STAT3, STAT5a, and STAT5b have a wide activation profile. STAT1 is activated by many different ligands including IFN family (IFN-Alpha, IFN-Beta, IFN-gamma and IL-10), gp130 family (IL-6, IL-11, LIF, CNTF, and G-CSF), and receptor tyrosine kinases (EGF, PDGF, and CSF-1). Two alternatively spliced transcript variants encoding distinct isoforms of STAT1 have been described.Specifications
P42224 | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
CANDF7,IMD31A,IMD31B,IMD31C,ISGF-3,STAT91 | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
1 hr. 10 min. |
Biotin | |
Cell Lysates | |
ELISA | |
6772 | |
Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal | |
96 Tests | |
Cell Lysate,20 μL | |
Signal transducer and activator of transcription 1-alpha/beta | |
1 hr. 20 min. |
For Research Use Only