Learn More
Thermo Scientific™ Phire Buffers
Choose from three formats of these buffers to optimize PCR using Thermo Scientific™ Phire™ Hot Start II DNA Polymerase.
£21.77 - £24.64
Product Code | Brand | For Use With (Application) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Quantity | Price | Quantity & Availability | ||||
10525709 |
Thermo Scientific™
F525L |
PCR, High-Fidelity PCR, High Performance PCR | 4 x 1.5mL |
£21.77 Pack of 4 |
Estimated Shipment date from Supplier 23-12-2019 |
Please sign in to purchase this item. Need a web account? Register with us today! | 10165847 |
Thermo Scientific™
F524L |
Phire DNA Polymerases. | 4 x 1.5mL |
£24.64 Pack of 4 |
EU Stock 21 | Please sign in to purchase this item. Need a web account? Register with us today! |
Description
Includes:
Detergent-free 5x Phire Reaction Buffer, 50mM MgCl2 solution, 100% DMSO
Recommended for:
PCR, High-Fidelity PCR, High Performance PCR