Learn More
Thermo Scientific™ Phire Reaction Buffer, detergent-free
Thermo Scientific™ Phire Reaction Buffer, Detergent-free, is recommended for PCR applications.
Brand: Thermo Scientific™ F525L
19.36 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 87
Additional Details : Weight : 0.02000kg
Description
Perform PCR applications with Thermo Scientific™ Phire™ Detergent-free Reaction Buffer.
Includes:
Detergent-free 5x Phire Reaction Buffer, 50mM MgCl2 solution, 100% DMSO
Recommended for:
PCR, High-Fidelity PCR, High Performance PCR
Specifications
Contains: • 4 x 1.5 mL 5X Phire Reaction Buffer, detergent-free (1X buffer provides 1.5 mM MgCl2) • 50 mM MgCl2 • DMSO Store at -20°C. |
|
Phire | |
Standard PCR | |
1.5 mL |
Phire Hot Start II DNA polymerase | |
4 x 1.5 mL | |
5X |
For Research Use Only. Not for use in diagnostic procedures.