Learn More
Liofilchem™ Contact Slide 2
Rose Bengal CAF Agar and Neutralizing / PCA and TTC and Neutralizing Flex Dip-slide with a selective medium for detection of yeasts and moulds and a non selective medium for total bacterial count.
Brand: Liofilchem™ 525272
Additional Details : Weight : 0.54900kg
Description
- Contact Slide 2 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of yeasts and moulds . The other medium is used for enumeration of bacteria.
Specifications
Bacteria, Yeast, Fungus | |
BD DIFCO | |
Agar | |
Non-sterile | |
Incubator | |
20 Pack |
Screw | |
Italy | |
Agar | |
Eukaryotic Microorganism, Fungi, Gram-positive Organisms, Fungi | |
10 and 25°C |